Assay Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024264.4 | NP_077226.2 | 1 - 2 | 313 | 81 | |
| GenBank mRNA | AK004977.1 | - | 1 - 2 | 314 | 81 | |
| BC055028.1 | - | 1 - 2 | 291 | 5098366 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27a1
Genomic Map