Assay Details
| Gene Symbol: Dnaic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175138.4 | NP_780347.2 | 13 - 14 | 1517 | 59 | |
| GenBank mRNA | AK030213.1 | - | 13 - 14 | 1451 | 59 | |
| AK047808.1 | - | 13 - 14 | 1518 | 59 | ||
| BC138785.1 | - | 13 - 14 | 1462 | 8861807 | 59 | |
| BC138787.1 | - | 13 - 14 | 1462 | 8861809 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaic1
Genomic Map