Assay Details
| Gene Symbol: Dnaic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175138.4 | NP_780347.2 | 1 - 2 | 226 | 61 | |
| GenBank mRNA | AK030213.1 | - | 1 - 2 | 160 | 61 | |
| AK047808.1 | - | 1 - 2 | 227 | 61 | ||
| BC138785.1 | - | 1 - 2 | 171 | 8861807 | 61 | |
| BC138787.1 | - | 1 - 2 | 171 | 8861809 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK030213.1,AK047808.1,BC138785.1,BC138787.1,NM_175138.4 transcripts.
Set Membership: |
|

Dnaic1
Genomic Map