Assay Details
| Gene Symbol: Hectd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175244.3 | NP_780453.1 | 10 - 11 | 1556 | 85 | |
| GenBank mRNA | AK051385.1 | - | 10 - 11 | 1569 | 85 | |
| AK139305.1 | - | 10 - 11 | 1584 | 85 | ||
| AK154378.1 | - | 10 - 11 | 1588 | 85 | ||
| AK155290.1 | - | 10 - 11 | 1569 | 85 | ||
| AK155676.1 | - | 10 - 11 | 1569 | 85 | ||
| AK163323.1 | - | 10 - 11 | 1591 | 85 | ||
| AK165482.1 | - | 10 - 11 | 1588 | 85 | ||
| AK170930.1 | - | 10 - 11 | 1569 | 85 | ||
| AK204023.1 | - | 1 - 2 | 67 | 85 | ||
| BC070411.1 | - | 10 - 11 | 1556 | 5706209 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hectd3
Genomic Map