Assay Details
| Gene Symbol: Hectd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175244.3 | NP_780453.1 | 17 - 18 | 2321 | 62 | |
| GenBank mRNA | AK051385.1 | - | 17 - 18 | 2334 | 62 | |
| AK139305.1 | - | 17 - 18 | 2349 | 62 | ||
| AK154378.1 | - | 17 - 18 | 2353 | 62 | ||
| AK155290.1 | - | 17 - 18 | 2334 | 62 | ||
| AK155676.1 | - | 17 - 18 | 2334 | 62 | ||
| AK163323.1 | - | 17 - 18 | 2356 | 62 | ||
| AK165482.1 | - | 17 - 18 | 2353 | 62 | ||
| AK170930.1 | - | 17 - 18 | 2334 | 62 | ||
| BC024415.1 | - | 3 - 4 | 389 | 62 | ||
| BC070411.1 | - | 17 - 18 | 2321 | 5706209 | 62 | |
| BC082276.1 | - | 7 - 8 | 775 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hectd3
Genomic Map