Assay Details
| Gene Symbol: Sec24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290785.1 | NP_001277714.1 | 21 - 22 | 3710 | 83 | |
| NM_175255.3 | NP_780464.2 | 21 - 22 | 3713 | 83 | ||
| GenBank mRNA | AK029139.1 | - | 21 - 22 | 3466 | 83 | |
| AK155182.1 | - | 21 - 22 | 3499 | 83 | ||
| BC117905.1 | - | 21 - 22 | 2977 | 83 | ||
| BC117906.1 | - | 5 - 6 | 532 | 83 | ||
| BG065014.2 | - | 2 - 3 | 83 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sec24a
Genomic Map