Assay Details
| Gene Symbol: Six3os1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015387.2 | 3 - 4 | 841 | 99 | |||
| GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 - 3 | 2350 | 99 | |
| AY589791.1 | - | 3 - 4 | 810 | 99 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145159 | AK053722.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145159 | BC065087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145159 | AY589790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145159 | NR_015386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145159 | AK021382.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145164 | AY589792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145164 | BC065087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145164 | NR_015385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145154 | AY589791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145154 | NR_015387.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93746 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163177 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159532 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145154 | AY589791.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145154 | NR_015387.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145164 | AY589792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145164 | BC065087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145164 | NR_015385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145159 | AK053722.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145159 | BC065087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145159 | AY589790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145159 | NR_015386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145159 | AK021382.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13260446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13521393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13260445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13260447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13063872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13480357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13541000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13511321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31411637 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10453393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453392 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431168_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10453390 | - | - |
| Mouse Genome 430 2.0 Array | 1431168_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431168_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700024189.mm.1 | NR_015387.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024189.mm.1 | NR_015387.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024189.mm.1 | TC1700002661.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431168_at | - | - |
| Mouse Expression Set 430 | 1431168_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431168_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431168_PM_at | - | - |

Six3os1
Genomic Map