| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600021654.mm.1 |
NM_175313.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021654.mm.1 |
NM_175313.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021654.mm.1 |
TC0600002796.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600021655.mm.1 |
NM_175313.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021655.mm.1 |
NM_175313.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021655.mm.1 |
TC0600002796.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600021651.mm.1 |
NM_175313.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021651.mm.1 |
NM_175313.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021651.mm.1 |
TC0600002796.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600021650.mm.1 |
NM_175313.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021650.mm.1 |
NM_175313.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021650.mm.1 |
TC0600002796.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600021653.mm.1 |
NM_175313.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021653.mm.1 |
NM_175313.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600021653.mm.1 |
TC0600002796.mm.1 |
MTA TC |