Assay Details
| Gene Symbol: Dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176835.2 | NP_789805.1 | 2 - 3 | 1071 | 99 | |
| GenBank mRNA | AK013323.1 | - | 2 - 3 | 1094 | 99 | |
| AK050294.1 | - | 2 - 3 | 1050 | 99 | ||
| BC039633.1 | - | 2 - 3 | 1071 | 5136672 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478229 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612379 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177075 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30478249 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03112673 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177075 | - | - |
| Whole Mouse Genome G4122A | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | BC039633.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK013323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK050294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | AK169115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257743 | NM_176835.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133962 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12079964 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12079963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12079965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12079966 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12230091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12079969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30478229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30478204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12335361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12230088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12230096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12390733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12017972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12390727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12208608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30478290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12390730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426686 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429773_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460468_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426685 | - | - |
| Mouse Genome 430 2.0 Array | 1460468_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1460468_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429773_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429773_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72778_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72778_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500007823.mm.1 | NM_176835.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500007823.mm.1 | NM_176835.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500007823.mm.1 | TC1500001007.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460468_s_at | - | - |
| Mouse Expression Set 430 | 1429773_at | - | - |
| Mouse Expression Set 430 | 1429773_PM_at | - | - |
| Mouse Expression Set 430 | 1460468_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460468_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460468_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429773_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429773_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712269 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712269 | - | - |
| mouse-6 v1.1 (Illumina) | scl072778.3_56-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072778.3_56-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450050 | - | - |

Dnajc22
Genomic Map