Assay Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164531.1 | NP_001158003.1 | 10 - 11 | 1183 | 65 | |
| NM_177319.3 | NP_796293.2 | 11 - 12 | 1321 | 65 | ||
| GenBank mRNA | AK159065.1 | - | 10 - 11 | 1157 | 65 | |
| BC024801.1 | - | 11 - 12 | 1299 | 65 | ||
| BC042595.1 | - | 11 - 12 | 1267 | 5363403 | 65 | |
More Information
Set Membership: |
|

Zfyve27
Genomic Map