Assay Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164531.1 | NP_001158003.1 | 5 - 6 | 663 | 70 | |
| NM_177319.3 | NP_796293.2 | 5 - 6 | 780 | 70 | ||
| GenBank mRNA | AK159065.1 | - | 5 - 6 | 637 | 70 | |
| AK199316.1 | - | 1 - 2 | 69 | |||
| BC024801.1 | - | 5 - 6 | 754 | 70 | ||
| BC042595.1 | - | 5 - 6 | 726 | 5363403 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve27
Genomic Map