Assay Details
| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177321.2 | NP_796295.1 | 2 - 3 | 252 | 78 | |
| GenBank mRNA | AF390177.2 | - | 2 - 3 | 252 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92700 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127896 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122878 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28470220 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122878 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127896 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07635245 | - | - |
| Whole Mouse Genome G4122A | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168319 | NM_177321.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P168319 | AF390177.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116081 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115629 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07635238 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122878 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10309140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10309136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10309139 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28470277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10174149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10118241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10174150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09766631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09799908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09813095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10412213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09985887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10166608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09766628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09967081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28470228 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395929 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451964_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395925 | - | - |
| Mouse Genome 430 2.0 Array | 1451964_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451964_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 338320_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 338320_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003485.mm.1 | NM_177321.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003485.mm.1 | NM_177321.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003485.mm.1 | TC1200000502.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003484.mm.1 | NM_177321.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003484.mm.1 | NM_177321.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003484.mm.1 | TC1200000502.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003486.mm.1 | NM_177321.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003486.mm.1 | NM_177321.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003486.mm.1 | TC1200000502.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451964_PM_at | - | - |
| Mouse Expression Set 430 | 1451964_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451964_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451964_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451964_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451964_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 832268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3130181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130181 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694320 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694320 | - | - |
| mouse-6 v1.1 (Illumina) | scl43181.4.1_246-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43181.4.1_246-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130181 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694320 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694320 | - | - |

Mia2
Genomic Map