Assay Details
| Gene Symbol: Rint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177323.3 | NP_796297.2 | 8 - 9 | 1207 | 117 | |
| GenBank mRNA | AK028707.1 | - | 7 - 8 | 1064 | 117 | |
| AK036805.1 | - | 8 - 9 | 1207 | 117 | ||
| AK135334.1 | - | 8 - 9 | 1228 | 117 | ||
| AK144144.1 | - | 8 - 9 | 1229 | 118 | ||
| AK193380.1 | - | 1 - 2 | 127 | 117 | ||
| BC065059.1 | - | 8 - 9 | 1207 | 6401664 | 117 | |
| BC145267.1 | - | 7 - 8 | 1023 | 9053821 | 117 | |
| BC150745.1 | - | 7 - 8 | 1023 | 9087656 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rint1
Genomic Map