Assay Details
| Gene Symbol: Slc9a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177353.3 | NP_796327.1 | 5 - 6 | 875 | 151 | |
| GenBank mRNA | AK041169.1 | - | 5 - 6 | 875 | 151 | |
| AK078577.1 | - | 5 - 6 | 920 | 151 | ||
| BC058750.1 | - | 5 - 6 | 881 | 6390297 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a7
Genomic Map