Assay Details
| Gene Symbol: Cyp2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110408.1 | 2 - 3 | 285 | 78 | ||
| GenBank mRNA | AK050453.1 | - | 2 - 3 | 275 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050453.1 transcripts.
Set Membership: |
|

Cyp2r1
Genomic Map