Assay Details
| Gene Symbol: Slc41a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177388.3 | NP_796362.1 | 7 - 8 | 1414 | 75 | |
| GenBank mRNA | AK038549.1 | - | 7 - 8 | 1415 | 75 | |
| BC138146.1 | - | 6 - 7 | 1252 | 8861165 | 75 | |
| BC138147.1 | - | 6 - 7 | 1252 | 8861166 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc41a2
Genomic Map