Assay Details
| Gene Symbol: Psd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177611.3 | NP_808279.1 | 10 - 11 | 2202 | 63 | |
| GenBank mRNA | AK043669.1 | - | 11 - 12 | 2494 | 63 | |
| AK150238.1 | - | 10 - 11 | 2231 | 63 | ||
| AK155249.1 | - | 10 - 11 | 2232 | 63 | ||
| AK163628.1 | - | 6 - 7 | 581 | 63 | ||
| AK169859.1 | - | 10 - 11 | 2231 | 63 | ||
| BC046518.1 | - | 2 - 3 | 129 | 63 | ||
| BC068231.1 | - | 10 - 11 | 2202 | 30361859 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd4
Genomic Map