Assay Details
| Gene Symbol: Psd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177611.3 | NP_808279.1 | 7 - 8 | 1939 | 71 | |
| GenBank mRNA | AK043669.1 | - | 8 - 9 | 2231 | 71 | |
| AK150238.1 | - | 7 - 8 | 1968 | 71 | ||
| AK155249.1 | - | 7 - 8 | 1969 | 71 | ||
| AK163628.1 | - | 3 - 4 | 318 | 71 | ||
| AK169859.1 | - | 7 - 8 | 1968 | 71 | ||
| BC068231.1 | - | 7 - 8 | 1939 | 30361859 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd4
Genomic Map