Assay Details
| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177624.3 | NP_808292.1 | 2 - 3 | 168 | 137 | |
| GenBank mRNA | AK040281.1 | - | 2 - 3 | 169 | 137 | |
| BC126945.1 | - | 2 - 3 | 169 | 40090204 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sntn
Genomic Map