Assay Details
| Gene Symbol: Cdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177647.4 | NP_808315.1 | 2 - 3 | 307 | 73 | |
| GenBank mRNA | AK034009.1 | - | 2 - 3 | 309 | 73 | |
| BC110560.2 | - | 2 - 3 | 295 | 40049634 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20962888 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402546 | AK087735.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110278 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155094 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142298 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20962922 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142298 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04411261 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155094 | - | - |
| Whole Mouse Genome G4122A | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479645 | NM_177647.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479645 | AK034009.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131461 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155094 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01352721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01352715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01352748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20962888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20963021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20962992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01272758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01171042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01414559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01230803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01230801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01352748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01665084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01242013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01188785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20963006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02015125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01665083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01947933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20962934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01845051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01947929 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468979 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468974 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200000112.mm.1 | NM_177647.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000112.mm.1 | NM_177647.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000112.mm.1 | TC0200000009.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200000111.mm.1 | NM_177647.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000111.mm.1 | NM_177647.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000111.mm.1 | TC0200000009.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200000109.mm.1 | NM_177647.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000109.mm.1 | NM_177647.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200000109.mm.1 | TC0200000009.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 429374 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1940603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721460 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721460 | - | - |
| mouse-6 v1.1 (Illumina) | scl21338.5_363-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21338.5_363-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721460 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721460 | - | - |

Cdnf
Genomic Map