Assay Details
| Gene Symbol: Cidec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301295.1 | NP_001288224.1 | 5 - 6 | 629 | 78 | |
| NM_178373.3 | NP_848460.1 | 5 - 6 | 613 | 78 | ||
| GenBank mRNA | AK080133.1 | - | 5 - 6 | 615 | 78 | |
| BC099676.1 | - | 4 - 5 | 585 | 5043221 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759513 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969131 | BC099676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969131 | NM_178373.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969131 | AK080133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969128 | BC099676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969128 | M61737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969128 | NM_178373.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969128 | AK080133.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10345 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156796 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969131 | BC099676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969131 | NM_178373.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969131 | AK080133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969128 | BC099676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969128 | M61737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969128 | NM_178373.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969128 | AK080133.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490456 | BC099676.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490456 | M61737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490456 | NM_178373.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490456 | AK080133.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06030968 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156796 | - | - |
| Whole Mouse Genome G4122A | A_51_P490456 | BC099676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490456 | M61737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490456 | NM_178373.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490456 | AK080133.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107444 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124864 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156796 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06030968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05745071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05211429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05631175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05211428 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05768677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05386755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05768683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05745072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05768678 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05625343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05689072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05142797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05416847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05629272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05825781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05493764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05723567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05629275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05493762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05614861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05214419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05629274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05719245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05055349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05503828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05723570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05765174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05055348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05493763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05508823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05768672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10546930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546932 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452260_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10546929 | - | - |
| Mouse Genome 430 2.0 Array | 1452260_at | - | - |
| Mouse Genome 430 2.0 Array | 1452260_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14311_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14311_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa11970_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600022440.mm.1 | NM_178373.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022440.mm.1 | NM_178373.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022440.mm.1 | TC0600002920.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600022441.mm.1 | NM_178373.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022441.mm.1 | NM_178373.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022441.mm.1 | TC0600002920.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452260_at | - | - |
| Mouse Expression Set 430 | 1452260_PM_at | - | - |
| Murine Genome U74v2 Set | 102016_PM_at | - | - |
| Murine Genome U74v2 Set | 102016_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452260_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452260_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452260_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452260_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 843553 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3450372 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3450372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222679 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222679 | - | - |
| mouse-6 v1.1 (Illumina) | scl28542.7_206-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450372 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28542.7_206-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1222679 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222679 | - | - |

Cidec
Genomic Map