Assay Details
| Gene Symbol: Cox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178379.3 | NP_848466.1 | 2 - 3 | 267 | 58 | |
| GenBank mRNA | AK010385.1 | - | 2 - 3 | 261 | 58 | |
| AK149999.1 | - | 2 - 3 | 257 | 58 | ||
| AK154668.1 | - | 2 - 3 | 266 | 58 | ||
| AK156865.1 | - | 2 - 3 | 266 | 58 | ||
| AK172718.1 | - | 2 - 3 | 266 | 58 | ||
| AK178239.1 | - | 1 - 2 | 62 | 58 | ||
| AK179799.1 | - | 1 - 2 | 62 | 58 | ||
| AK189355.1 | - | 1 - 2 | 62 | 58 | ||
| AK199666.1 | - | 1 - 2 | 62 | 58 | ||
| AK207639.1 | - | 1 - 2 | 62 | 58 | ||
| AK210222.1 | - | 1 - 2 | 62 | 58 | ||
| AK212782.1 | - | 1 - 2 | 62 | 58 | ||
| AK212812.1 | - | 1 - 2 | 62 | 58 | ||
| AK218286.1 | - | 1 - 2 | 62 | 58 | ||
| BC018301.1 | - | 2 - 3 | 219 | 58 | ||
| BC038046.1 | - | 2 - 3 | 220 | 4206826 | 58 | |
| BY078593.1 | - | 2 - 3 | 267 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox10
Genomic Map