Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 21 - 22 | 3117 | 104 | |
| GenBank mRNA | AK137602.1 | - | 21 - 22 | 3053 | 104 | |
| AK147331.1 | - | 21 - 22 | 3049 | 104 | ||
| AK148775.1 | - | 20 - 21 | 3239 | 104 | ||
| AK161840.1 | - | 21 - 22 | 3049 | 104 | ||
| AK164509.1 | - | 21 - 22 | 3053 | 104 | ||
| AK168703.1 | - | 21 - 22 | 3049 | 104 | ||
| BC025807.1 | - | 10 - 11 | 1345 | 104 | ||
| BC036127.1 | - | 21 - 22 | 3041 | 4955003 | 104 | |
| BC041774.1 | - | 18 - 19 | 2767 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a2
Genomic Map