Assay Details
| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178593.3 | NP_848708.2 | 3 - 4 | 401 | 103 | |
| GenBank mRNA | AK133961.1 | - | 3 - 4 | 356 | 103 | |
| AK158429.1 | - | 3 - 4 | 342 | 103 | ||
| AK171914.1 | - | 3 - 4 | 324 | 103 | ||
| BC025872.1 | - | 3 - 4 | 321 | 5037953 | 103 | |
More Information
Set Membership: |
|

Rcsd1
Genomic Map