Assay Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178599.4 | NP_848714.1 | 1 - 2 | 108 | 75 | |
| GenBank mRNA | AK012653.1 | - | 1 - 2 | 94 | 75 | |
| AK013015.1 | - | 1 - 2 | 79 | 75 | ||
| AK013371.1 | - | 1 - 2 | 81 | 75 | ||
| AK077934.1 | - | 1 - 2 | 108 | 75 | ||
| AK151560.1 | - | 1 - 2 | 91 | 75 | ||
| AK158078.1 | - | 1 - 2 | 91 | 75 | ||
| AK170011.1 | - | 1 - 2 | 81 | 75 | ||
| BC098183.1 | - | 1 - 2 | 84 | 5150466 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Commd8
Genomic Map