Assay Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163328.1 | NP_001156800.1 | 6 - 7 | 887 | 108 | |
| NM_001163329.1 | NP_001156801.1 | 5 - 6 | 480 | 108 | ||
| NM_001163330.1 | NP_001156802.1 | 5 - 6 | 480 | 108 | ||
| NM_178631.4 | NP_848746.1 | 7 - 8 | 980 | 108 | ||
| GenBank mRNA | AK038465.1 | - | 5 - 6 | 458 | 108 | |
| AK079659.1 | - | 6 - 7 | 1709 | 108 | ||
| BC052358.1 | - | 5 - 6 | 441 | 6534677 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ralyl
Genomic Map