Assay Details
| Gene Symbol: Pkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178654.4 | NP_848769.2 | 19 - 20 | 2909 | 139 | |
| GenBank mRNA | AK049713.1 | - | 19 - 20 | 2896 | 139 | |
| AK054281.1 | - | 5 - 6 | 513 | 139 | ||
| AK083425.1 | - | 14 - 15 | 1707 | 139 | ||
| AK133298.1 | - | 19 - 20 | 2898 | 139 | ||
| AK171092.1 | - | 19 - 20 | 2840 | 139 | ||
| BC052073.1 | - | 20 - 21 | 2832 | 6400440 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkn2
Genomic Map