Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178664.5 | NP_848779.2 | 9 - 10 | 970 | 86 | |
| GenBank mRNA | AK031361.1 | - | 9 - 10 | 86 | ||
| AK156318.1 | - | 9 - 10 | 948 | 86 | ||
| BC022739.1 | - | 3 - 4 | 282 | 86 | ||
| BC062106.1 | - | 3 - 4 | 829 | 86 | ||
| BC088993.1 | - | 9 - 10 | 835 | 6416623 | 86 | |
| BC125626.1 | - | 9 - 10 | 943 | 40130141 | 86 | |
| BC132193.1 | - | 9 - 10 | 943 | 40130470 | 86 | |
| BC145163.1 | - | 9 - 10 | 947 | 86 | ||
| BC145165.1 | - | 9 - 10 | 1009 | 86 | ||
| BC145166.1 | - | 9 - 10 | 943 | 9053717 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gntl1
Genomic Map