Assay Details
| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178693.4 | NP_848808.1 | 2 - 3 | 230 | 65 | |
| GenBank mRNA | AK039422.1 | - | 2 - 3 | 231 | 65 | |
| AK039676.1 | - | 2 - 3 | 231 | 65 | ||
| AK043574.1 | - | 2 - 3 | 206 | 65 | ||
| AK050459.1 | - | 2 - 3 | 240 | 65 | ||
| AK163270.1 | - | 999 | 65 | |||
| BC139373.1 | - | 2 - 3 | 273 | 8862395 | 65 | |
| BC139376.1 | - | 2 - 3 | 270 | 8862398 | 65 | |
| BC145615.1 | - | 2 - 3 | 273 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Coq4
Genomic Map