Assay Details
| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178693.4 | NP_848808.1 | 4 - 5 | 423 | 69 | |
| GenBank mRNA | AK039422.1 | - | 4 - 5 | 424 | 69 | |
| AK039676.1 | - | 4 - 5 | 424 | 69 | ||
| AK043574.1 | - | 4 - 5 | 399 | 69 | ||
| AK050459.1 | - | 4 - 5 | 433 | 69 | ||
| BC139373.1 | - | 4 - 5 | 466 | 8862395 | 69 | |
| BC139376.1 | - | 4 - 5 | 463 | 8862398 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq4
Genomic Map