Assay Details
| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178693.4 | NP_848808.1 | 6 - 7 | 653 | 71 | |
| GenBank mRNA | AK039422.1 | - | 6 - 7 | 654 | 71 | |
| AK039676.1 | - | 6 - 7 | 654 | 71 | ||
| AK043574.1 | - | 6 - 7 | 629 | 71 | ||
| AK050459.1 | - | 6 - 7 | 663 | 71 | ||
| AK163270.1 | - | 1499 | 71 | |||
| BC039570.1 | - | 1 - 2 | 301 | 71 | ||
| BC139373.1 | - | 6 - 7 | 696 | 8862395 | 71 | |
| BC139376.1 | - | 6 - 7 | 693 | 8862398 | 71 | |
| BC145615.1 | - | 6 - 7 | 667 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099461 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099456 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317797 | AK033897.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148781 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146218 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461219 | AK163270.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461219 | AK039422.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461219 | NM_178693.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21099452 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146218 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00333085 | - | - |
| Whole Mouse Genome G4122A | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK050459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK163270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | BC039570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK039422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P486045 | AK039676.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461219 | AK163270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461219 | AK039422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461219 | NM_178693.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100382 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02254945 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01238382 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02254950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01238387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02254948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099461 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21099455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21099517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01296995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01297003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01192592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01296994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02254948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02254945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01296996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01296999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01297001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01192591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02254950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01192593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01815745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01296991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01297004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21099414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470748 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470750 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435231_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470741 | - | - |
| Mouse Genome 430 2.0 Array | 1435231_at | - | - |
| Mouse Genome 430 2.0 Array | 1435231_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227683_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227683_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200003740.mm.1 | NM_178693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003740.mm.1 | TC0200000510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200003742.mm.1 | NM_178693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003742.mm.1 | TC0200000510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200003741.mm.1 | NM_178693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003741.mm.1 | TC0200000510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200003739.mm.1 | NM_178693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003739.mm.1 | TC0200000510.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435231_at | - | - |
| Mouse Expression Set 430 | 1435231_PM_at | - | - |
| Murine Genome U74v2 Set | 92261_PM_at | - | - |
| Murine Genome U74v2 Set | 161675_f_at | - | - |
| Murine Genome U74v2 Set | 92261_at | - | - |
| Murine Genome U74v2 Set | 161675_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435231_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435231_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435231_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435231_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1660551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2647972 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2647972 | - | - |
| mouse-6 v1.1 (Illumina) | scl21111.7_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21111.7_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2647972 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2885590 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2885590 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2647972 | - | - |

Coq4
Genomic Map