Assay Details
| Gene Symbol: Grsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098476.2 | NP_001091946.1 | 4 - 5 | 541 | 136 | |
| NM_178700.5 | NP_848815.2 | 4 - 5 | 869 | 136 | ||
| GenBank mRNA | AK044889.1 | - | 4 - 5 | 870 | 136 | |
| AK045986.1 | - | 4 - 5 | 823 | 136 | ||
| AK077782.1 | - | 4 - 5 | 870 | 136 | ||
| AK089014.1 | - | 3 - 4 | 405 | 136 | ||
| AK168385.1 | - | 4 - 5 | 512 | 136 | ||
| AK168459.1 | - | 4 - 5 | 512 | 136 | ||
| AK210897.1 | - | 1 - 2 | 56 | 136 | ||
| AK218546.1 | - | 1 - 2 | 56 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grsf1
Genomic Map