Assay Details
| Gene Symbol: Pcid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178708.3 | NP_848823.2 | 11 - 12 | 1394 | 74 | |
| GenBank mRNA | AK042945.1 | - | 11 - 12 | 884 | 74 | |
| AK045796.1 | - | 11 - 12 | 874 | 74 | ||
| AK048722.1 | - | 11 - 12 | 885 | 74 | ||
| AK082891.1 | - | 11 - 12 | 1460 | 74 | ||
| AK163697.1 | - | 11 - 12 | 881 | 74 | ||
| BC006047.1 | - | 11 - 12 | 883 | 74 | ||
| BC072621.1 | - | 8 - 9 | 1328 | 74 | ||
| BC118533.1 | - | 11 - 12 | 1394 | 40095328 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcid2
Genomic Map