Assay Details
| Gene Symbol: Rxfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178717.3 | NP_848832.1 | 1 | 2252 | 133 | |
| GenBank mRNA | AK043414.1 | - | 1 | 2277 | 133 | |
| AK046367.1 | - | 1 | 2003 | 133 | ||
| AK084001.1 | - | 1 | 2254 | 133 | ||
| BC053073.1 | - | 1 | 1674 | 5686363 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytokinesis
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
signal transducer activity
G-protein coupled receptor activity
G-protein coupled peptide receptor activity
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytokinesis
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
signal transducer activity
G-protein coupled receptor activity
G-protein coupled peptide receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013955 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013952 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013954 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013956 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054832 | BC053073.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054832 | AK046367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054832 | AK084001.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054832 | AY170825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054832 | AY633762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054827 | AK084001.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054827 | NM_178717.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539888 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148750 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054832 | BC053073.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054832 | AK046367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054832 | AK084001.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054832 | AY170825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054832 | AY633762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054827 | AK084001.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054827 | NM_178717.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P255406 | BC053073.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P255406 | AK046367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P255406 | AK084001.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P255406 | AK043414.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P255406 | NM_178717.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30013954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30014009 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148750 | - | - |
| Whole Mouse Genome G4122A | A_52_P255406 | BC053073.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P255406 | AK046367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P255406 | AK084001.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P255406 | AK043414.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P255406 | NM_178717.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116543 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111109 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142594 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142585 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12142586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013956 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013954 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013955 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30013952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30014004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30013925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11917615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11985019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11839059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11903993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11917613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30014019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12214099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30013925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10427770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427768 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10427766 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500009768.mm.1 | NM_178717.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009768.mm.1 | NM_178717.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500009768.mm.1 | TC1500001212.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 137146_at | - | - |
| Murine Genome U74v2 Set | 137146_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5550022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240943 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240943 | - | - |
| mouse-6 v1.1 (Illumina) | scl47391.1.22_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47391.1.22_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240943 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240943 | - | - |

Rxfp3
Genomic Map