Assay Details
| Gene Symbol: Tnfrsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178931.2 | NP_849262.1 | 4 - 5 | 489 | 97 | |
| GenBank mRNA | AF515707.1 | - | 4 - 5 | 458 | 97 | |
| AY264405.1 | - | 4 - 5 | 489 | 97 | ||
| BC022125.1 | - | 2 - 3 | 220 | 97 | ||
| BC104054.1 | - | 4 - 5 | 489 | 40040309 | 97 | |
| BC104055.1 | - | 4 - 5 | 489 | 40040311 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf14
Genomic Map