Assay Details
| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181400.3 | NP_852065.2 | 12 - 13 | 2602 | 69 | |
| GenBank mRNA | AK028113.1 | - | 6 - 7 | 993 | 69 | |
| BC040337.1 | - | 12 - 13 | 2463 | 3586908 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr47
Genomic Map