Assay Details
| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181400.3 | NP_852065.2 | 2 - 3 | 490 | 88 | |
| GenBank mRNA | AK047357.1 | - | 2 - 3 | 490 | 88 | |
| AK163422.1 | - | 2 - 3 | 381 | 88 | ||
| BC040337.1 | - | 2 - 3 | 351 | 3586908 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr47
Genomic Map