Assay Details
| Gene Symbol: Supv3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181423.2 | NP_852088.1 | 12 - 13 | 1603 | 69 | |
| GenBank mRNA | AJ968954.1 | - | 12 - 13 | 1630 | 69 | |
| AK031911.1 | - | 12 - 13 | 1610 | 69 | ||
| AK143755.1 | - | 12 - 13 | 1628 | 69 | ||
| BC049796.1 | - | 12 - 13 | 1603 | 6332552 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supv3l1
Genomic Map