Assay Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024282.3 | NP_077244.1 | 1 - 2 | 532 | 125 | |
| GenBank mRNA | AK020014.1 | - | 1 - 2 | 417 | 125 | |
| AK049579.1 | - | 1 - 2 | 301 | 125 | ||
| AK146492.1 | - | 1 - 2 | 265 | 125 | ||
| AK160733.1 | - | 1 - 2 | 121 | 125 | ||
| BC002200.1 | - | 1 - 2 | 201 | 3489351 | 125 | |
| BC046816.1 | - | 1 - 2 | 239 | 5702476 | 125 | |
More Information
Set Membership: |
|

Desi2
Genomic Map