Assay Details
| Gene Symbol: Traf3ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134000.3 | NP_598761.2 | 3 - 4 | 1413 | 110 | |
| GenBank mRNA | AB238206.1 | - | 2 - 3 | 992 | 110 | |
| AB238207.1 | - | 2 - 3 | 992 | 110 | ||
| AK031556.1 | - | 3 - 4 | 1138 | 110 | ||
| AK037407.1 | - | 3 - 4 | 1220 | 110 | ||
| AK052190.1 | - | 3 - 4 | 1413 | 110 | ||
| AK098107.1 | - | 3 - 4 | 1135 | 110 | ||
| BC008534.1 | - | 2 - 3 | 569 | 110 | ||
| BC008535.1 | - | 2 - 3 | 569 | 110 | ||
| BC096483.1 | - | 3 - 4 | 1094 | 5310588 | 110 | |
| BC138234.1 | - | 3 - 4 | 1183 | 8861253 | 110 | |
| BC138235.1 | - | 3 - 4 | 1183 | 8861254 | 110 | |
| JX869941.1 | - | 3 - 4 | 1420 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip2
Genomic Map