Assay Details
| Gene Symbol: Suox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173733.3 | NP_776094.2 | 2 - 3 | 126 | 96 | |
| GenBank mRNA | AK149422.1 | - | 2 - 3 | 114 | 96 | |
| AK154608.1 | - | 2 - 3 | 114 | 96 | ||
| BC027197.1 | - | 1 - 2 | 81 | 4160277 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154608.1 transcripts.
Set Membership: |
|

Suox
Genomic Map