Assay Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173751.4 | NP_776112.1 | 8 - 9 | 1094 | 120 | |
| GenBank mRNA | AK030933.1 | - | 5 - 6 | 532 | 120 | |
| AK087956.1 | - | 7 - 8 | 968 | 120 | ||
| AK133553.1 | - | 7 - 8 | 1024 | 120 | ||
| AK138031.1 | - | 7 - 8 | 945 | 120 | ||
| AK143918.1 | - | 7 - 8 | 993 | 120 | ||
| AK146075.1 | - | 7 - 8 | 1022 | 120 | ||
| AK169899.1 | - | 7 - 8 | 1020 | 120 | ||
| BC052054.1 | - | 7 - 8 | 1428 | 120 | ||
| BC056459.1 | - | 8 - 9 | 1052 | 5718157 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilvbl
Genomic Map