Assay Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173751.4 | NP_776112.1 | 10 - 11 | 1362 | 84 | |
| GenBank mRNA | AK030933.1 | - | 7 - 8 | 800 | 84 | |
| AK087956.1 | - | 9 - 10 | 1236 | 84 | ||
| AK146075.1 | - | 9 - 10 | 1290 | 84 | ||
| AK169899.1 | - | 9 - 10 | 1288 | 84 | ||
| BC052054.1 | - | 9 - 10 | 1696 | 84 | ||
| BC056459.1 | - | 10 - 11 | 1320 | 5718157 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilvbl
Genomic Map