Assay Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173757.3 | NP_776118.2 | 1 - 2 | 71 | ||
| GenBank mRNA | AK076008.1 | - | 1 - 2 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps27
Genomic Map