Assay Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173757.3 | NP_776118.2 | 9 - 10 | 858 | 67 | |
| GenBank mRNA | AK076008.1 | - | 9 - 10 | 857 | 67 | |
| AK087686.1 | - | 9 - 10 | 855 | 67 | ||
| AK172922.1 | - | 2 - 3 | 2277 | 67 | ||
| BC049149.1 | - | 9 - 10 | 785 | 6309524 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps27
Genomic Map