Assay Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 11 - 12 | 1222 | 103 | |
| GenBank mRNA | AK030002.1 | - | 6 - 7 | 103 | ||
| BC096038.1 | - | 8 - 9 | 701 | 103 | ||
| BC110670.1 | - | 10 - 11 | 943 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapt1
Genomic Map