Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 9 - 10 | 1458 | 63 | |
| GenBank mRNA | AK050143.1 | - | 6 - 7 | 953 | 63 | |
| AK178410.1 | - | 1 - 2 | 132 | 63 | ||
| AK196619.1 | - | 1 - 2 | 132 | 63 | ||
| BC026405.1 | - | 9 - 10 | 1345 | 4211657 | 63 | |
| BC089586.1 | - | 6 - 7 | 962 | 63 | ||
| BC139196.1 | - | 6 - 7 | 846 | 8862218 | 63 | |
| BC139197.1 | - | 6 - 7 | 846 | 8862219 | 63 | |
| DQ205645.1 | - | 8 - 9 | 1171 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insc
Genomic Map