Assay Details
| Gene Symbol: Klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173780.4 | NP_776141.1 | 2 - 3 | 110 | 105 | |
| GenBank mRNA | AK044185.1 | - | 2 - 3 | 111 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044185.1, NM_173780.4 transcripts.
Set Membership: |
|

Klf8
Genomic Map