Assay Details
| Gene Symbol: E030024N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033228.1 | 1 | 1235 | 125 | ||
| GenBank mRNA | AK053172.1 | - | 1 | 1239 | 125 | |
| AK142495.1 | - | 1 | 1235 | 125 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E030024N20Rik
Genomic Map