Assay Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177024.4 | NP_795998.2 | 2 - 3 | 148 | 127 | |
| GenBank mRNA | AK036858.1 | - | 201 | 127 | ||
| AK036968.1 | - | 2 - 3 | 201 | 127 | ||
| AK137654.1 | - | 213 | 127 | |||
| BC115419.1 | - | 2 - 3 | 148 | 40093437 | 127 | |
| BC115420.2 | - | 2 - 3 | 148 | 40093438 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11b
Genomic Map